Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Phenylenediacetonitrile, 98%

Catalog No. AAA1314218
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1314218 50 g
AAA1314206 5 g
AAA1314214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1314218 Supplier Thermo Scientific Chemicals Supplier No. A1314218
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 613-73-0 | C10H8N2 | 156.19 g/mol

1,2-Phenylenediacetonitrile is used as API and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Phenylenediacetonitrile is used as API and intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-73-0
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00001905
InChI Key FWPFXBANOKKNBR-UHFFFAOYSA-N
Synonym 1,2-phenylenediacetonitrile, o-phenylenediacetonitrile, 1,2-bis cyanomethyl benzene, o-xylylene dicyanide, 2-2-cyanomethyl phenyl acetonitrile, o-benzenediacetonitrile, 2,2'-1,2-phenylene diacetonitrile, o-bis cyanomethyl benzene, 1,2-benzenediacetonitrile, o-benzendiacetonitrile
PubChem CID 69180
IUPAC Name 2-[2-(cyanomethyl)phenyl]acetonitrile
SMILES N#CCC1=CC=CC=C1CC#N

Specifications

Melting Point 57°C to 61°C
Quantity 50 g
UN Number UN3439
Beilstein 510075
Solubility Information Insoluble in water.
Formula Weight 156.19
Percent Purity 98%
Chemical Name or Material 1,2-Phenylenediacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 210-351-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.