Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Phenylenediamine Sulfate 97.0+%, TCI America™
SDP

Catalog No. P017325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P017325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P017325G Supplier TCI America Supplier No. P017325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74710-09-1
Molecular Formula C6H10N2O4S
Molecular Weight (g/mol) 206.216
MDL Number MFCD00054335
InChI Key URGXGBOBXYFSAF-UHFFFAOYSA-N
Synonym 1,2-Diaminobenzene Sulfate
PubChem CID 44630426
IUPAC Name benzene-1,2-diamine;sulfuric acid
SMILES C1=CC=C(C(=C1)N)N.OS(=O)(=O)O

Specifications

Quantity 25 g
UN Number 2811
Formula Weight 206.22
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Phenylenediamine Sulfate
EINECSNumber (3)-0185
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.