Learn More
Medchemexpress LLC 1,2-piperidinedicarboxylic acid, 1-(1,1-dimethylethyl) ester, (2S)- | 26250-84-0 | 229.28 | 1 G
SDP
Catalog No. 5000446785
Encompass_Preferred
This product is Boc-Pip-OH, also known as Boc-L-pipecolic acid, identified as 1,2-piperidinedicarboxylic acid, 1-(1,1-dimethylethyl) ester, (2S)-. This compound functions as a t-butyloxycarbonylamino acid and can serve as a radical precursor, making it suitable for various biochemical applications.
  • Used as a biochemical reagent.
  • Acts as a t-butyloxycarbonylamino acid.
  • Can serve as a radical precursor.
  • High purity for research applications.
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-004-46785 Supplier Medchemexpress LLC Supplier No. HYW0077431G
This product is Boc-Pip-OH, also known as Boc-L-pipecolic acid, identified as 1,2-piperidinedicarboxylic acid, 1-(1,1-dimethylethyl) ester, (2S)-. This compound functions as a t-butyloxycarbonylamino acid and can serve as a radical precursor, making it suitable for various biochemical applications.
  • Used as a biochemical reagent.
  • Acts as a t-butyloxycarbonylamino acid.
  • Can serve as a radical precursor.
  • High purity for research applications.
  • Available in multiple pack sizes for experimental flexibility.