Learn More
Medchemexpress LLC 1,2-Piperidinedicarboxylic acid, 4-hydroxy-, 1-(1,1-dimethylethyl) 2-methyl ester, (2S,4S)- | 254882-14-9 | 99.7% | 259.30 | 25 MG
SDP
Catalog No. 5000402307
Encompass_Preferred
AN-12-H5 intermediate-1 is an intermediate of the viral inhibitor AN-12-H5 and can be used to synthesize Antibody-Drug Conjugates (ADCs). As an antienterovirus compound, AN-12-H5 targets the replication processes of PV and enterovirus 71 (EV71), showing no inhibitory effect on PI4KB activity and only moderate inhibitory effects on PI 3-kinase activity.
  • Appearance: Viscous liquid
  • Color: Colorless to light yellow
Catalog No. 50-004-02307 Supplier Medchemexpress LLC Supplier No. 25MGHY22055B
Add to Cart
Add to Cart
AN-12-H5 intermediate-1 is an intermediate of the viral inhibitor AN-12-H5 and can be used to synthesize Antibody-Drug Conjugates (ADCs). As an antienterovirus compound, AN-12-H5 targets the replication processes of PV and enterovirus 71 (EV71), showing no inhibitory effect on PI4KB activity and only moderate inhibitory effects on PI 3-kinase activity.
  • Appearance: Viscous liquid
  • Color: Colorless to light yellow