Learn More
Medchemexpress LLC 1,2-Piperidinedicarboxylic acid, 4-hydroxy-, 1-(1,1-dimethylethyl) 2-methyl ester, (2S,4S)- | 254882-14-9 | 99.7% | 259.30 | 250 MG
SDP
Catalog No. 5000403243
Encompass_Preferred
AN-12-H5 intermediate-1 is an intermediate of the viral inhibitor AN-12-H5. It can be used to synthesize Antibody-Drug Conjugates (ADCs). AN-12-H5 is an antienterovirus compound that targets the replication processes of PV and enterovirus 71 (EV71). AN-12-H5 shows no inhibitory effect on PI4KB activity and only moderate inhibitory effects on PI 3-kinase activity.
  • Targets the replication processes of PV and enterovirus 71
  • Shows no inhibitory effect on PI4KB activity
  • Can be used to synthesize antibody-drug conjugates (ADCs)
Catalog No. 50-004-03243 Supplier Medchemexpress LLC Supplier No. 250MGHY22055B
AN-12-H5 intermediate-1 is an intermediate of the viral inhibitor AN-12-H5. It can be used to synthesize Antibody-Drug Conjugates (ADCs). AN-12-H5 is an antienterovirus compound that targets the replication processes of PV and enterovirus 71 (EV71). AN-12-H5 shows no inhibitory effect on PI4KB activity and only moderate inhibitory effects on PI 3-kinase activity.
  • Targets the replication processes of PV and enterovirus 71
  • Shows no inhibitory effect on PI4KB activity
  • Can be used to synthesize antibody-drug conjugates (ADCs)