Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-1,2-Propanediol

Catalog No. 54024225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-024-225G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 54-024-225G Supplier Sigma Aldrich Supplier No. 54024225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-1,2-Propanediol undergoes metabolization in the bovine rumen fluid by the rumen microbiota with the emission of sulfur-containing gases.Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Specifications

CAS 79831-76-8
Melting Point 213°C to 217°C
Molecular Formula C8H15NO4
Refractive Index n20/D 1.434
Linear Formula C8H15NO4
MDL Number MFCD00017555
Quantity 100 mg
Synonym (R)-(-)-Propylene glycerol; (R)-(-)-Propylene glycol
Molecular Weight (g/mol) 189.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.