Learn More
Medchemexpress LLC (±)-1,2-propanediol-d6 | 52910-80-2 | MFCD00142483 | 99.0% | 82.13 | C3H2D6O2 | 1mg
SDP
Catalog No. 5000269925
Encompass_Preferred
( )-1 2-Propanediol-d6 is the deuterium labeled ( )-1 2-Propanediol ( )-1 2-Propanediol (1 2-(RS)-Propanediol) is an aliphatic alcohol and frequently used as an excipient in many agent formulations to increase the solubility and stability of agents[1][2]
Catalog No. 50-002-69925 Supplier Medchemexpress LLC Supplier No. HYY0921S21MG
( )-1 2-Propanediol-d6 is the deuterium labeled ( )-1 2-Propanediol ( )-1 2-Propanediol (1 2-(RS)-Propanediol) is an aliphatic alcohol and frequently used as an excipient in many agent formulations to increase the solubility and stability of agents[1][2]