Learn More
Medchemexpress LLC (±)-1,2-Propanediol-d8 | 80156-55-4 | 99.6% | 84.14 | 250 MG
SDP
Catalog No. 5000366087
Encompass_Preferred
(±)-1,2-Propanediol is an aliphatic alcohol frequently utilized as an excipient in numerous molecular preparations. Its primary function as an excipient is to enhance the solubility and stability of the active molecules within formulations. It is also recognized as an important bulk chemical product.
  • Functions as an aliphatic alcohol.
  • Acts as an excipient in molecular preparations.
  • Increases solubility of active molecules.
  • Enhances stability of active molecules.
  • Can be used as an organic solvent.
  • Serves as a diluent in drug preparations.
  • Is an important bulk chemical product.
Catalog No. 50-003-66087 Supplier Medchemexpress LLC Supplier No. 250MGHYY0921S
(±)-1,2-Propanediol is an aliphatic alcohol frequently utilized as an excipient in numerous molecular preparations. Its primary function as an excipient is to enhance the solubility and stability of the active molecules within formulations. It is also recognized as an important bulk chemical product.
  • Functions as an aliphatic alcohol.
  • Acts as an excipient in molecular preparations.
  • Increases solubility of active molecules.
  • Enhances stability of active molecules.
  • Can be used as an organic solvent.
  • Serves as a diluent in drug preparations.
  • Is an important bulk chemical product.