Learn More
Medchemexpress LLC 1,2-propanediol-d8 | 80156-55-4 | MFCD00142484 | 99.6% | 84.14 g/mol | C3D8O2 | 50mg
SDP
Catalog No. 5000268649
Encompass_Preferred
( )-1 2-Propanediol-d8 is the deuterium labeled ( )-1 2-Propanediol ( )-1 2-Propanediol (1 2-(RS)-Propanediol) is an aliphatic alcohol and frequently used as an excipient in many agent formulations to increase the solubility and stability of agents[1][2]
Catalog No. 50-002-68649 Supplier Medchemexpress LLC Supplier No. HYY0921S50MG
( )-1 2-Propanediol-d8 is the deuterium labeled ( )-1 2-Propanediol ( )-1 2-Propanediol (1 2-(RS)-Propanediol) is an aliphatic alcohol and frequently used as an excipient in many agent formulations to increase the solubility and stability of agents[1][2]