Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Propanediol OR, Macron Fine Chemicals™

Catalog No. MK19253 Shop All Avantor Products
Change view
Click to view available options
Glass; 3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-1925-3 Glass; 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-1925-3 Supplier Avantor Supplier No. 192562

missing translation for 'validMainframeMsg'

Specifications

CAS 57-55-6
Molecular Formula C3H8O2
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
SMILES CC(CO)O
IUPAC Name propane-1,2-diol
Molecular Weight (g/mol) 76.095
PubChem CID 1030
ChEBI CHEBI:16997
Formula Weight 76.1
Quantity 3 kg
Packaging Glass
Chemical Name or Material 1, 2-Propanediol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.