Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-propylene-d6 carbonate | 202480-74-8 | MFCD00142486 | 98.7% | 108.13 | C4D6O3 | 10 MG
SDP

Catalog No. 5000301773
Encompass_Preferred

1,2-Propylene-d6 carbonate (CAS 202480-74-8) is the deuterium-labeled analogue of 1,2-propylene carbonate, supplied as a colorless to light yellow liquid for use as an isotopically labeled reagent or analytical standard in research applications such as NMR, isotopic tracing, and mass spectrometry.

  • Deuterium-labeled (d6) isotopic enrichment.
  • High purity (98.65%).
  • Molecular formula C4D6O3 and molecular weight 108.13.
  • Appearance: colorless to light yellow liquid.
  • Product page includes datasheet and safety data sheet for handling information.

Catalog No. 50-003-01773 Supplier Medchemexpress LLC Supplier No. HYW032349S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Propylene-d6 carbonate (CAS 202480-74-8) is the deuterium-labeled analogue of 1,2-propylene carbonate, supplied as a colorless to light yellow liquid for use as an isotopically labeled reagent or analytical standard in research applications such as NMR, isotopic tracing, and mass spectrometry.

  • Deuterium-labeled (d6) isotopic enrichment.
  • High purity (98.65%).
  • Molecular formula C4D6O3 and molecular weight 108.13.
  • Appearance: colorless to light yellow liquid.
  • Product page includes datasheet and safety data sheet for handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.