Learn More
Medchemexpress LLC 1,2-propylene-d6 carbonate | 202480-74-8 | MFCD00142486 | 98.7% | 108.13 | C4D6O3 | 100 MG
SDP
Catalog No. 5000299411
Encompass_Preferred
1,2-Propylene-d6 carbonate is the deuterium-labeled analogue of 1,2-propylene carbonate used as a stable-isotope tracer and internal standard in analytical techniques such as NMR, GC-MS, and LC-MS. It is provided with high isotopic and chemical purity for quantitative studies and metabolic tracing.
  • High isotopic labeling (d6) for clear mass shifts.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity for reliable quantitative analysis.
  • Available in small research-scale packaging for method development.
  • Molecular weight 108.13; formula C4D6O3.
Catalog No. 50-002-99411 Supplier Medchemexpress LLC Supplier No. HYW032349S100MG
1,2-Propylene-d6 carbonate is the deuterium-labeled analogue of 1,2-propylene carbonate used as a stable-isotope tracer and internal standard in analytical techniques such as NMR, GC-MS, and LC-MS. It is provided with high isotopic and chemical purity for quantitative studies and metabolic tracing.
  • High isotopic labeling (d6) for clear mass shifts.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity for reliable quantitative analysis.
  • Available in small research-scale packaging for method development.
  • Molecular weight 108.13; formula C4D6O3.