Learn More
Medchemexpress LLC 1,2-propylene-d6 carbonate | 202480-74-8 | MFCD00142486 | 98.7% | 108.13 | C4D6O3 | 50 MG
SDP
Catalog No. 5000299321
Encompass_Preferred
1,2-Propylene-d6 carbonate is the deuterium-labeled analogue of 1,2-propylene carbonate used as an analytical tracer and internal standard for quantitative NMR, GC-MS, and LC-MS. It is supplied as a high-purity, isotopically labeled small-molecule reagent for method development, calibration, and quantitation workflows.
  • Deuterium-labeled (d6) isotopologue for mass and NMR differentiation.
  • High purity (98.65%) suitable for analytical applications.
  • Molecular weight 108.13 and chemical formula C4D6O3.
  • Intended use as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Available in small package sizes for analytical and method development.
  • Compatible with quantitative workflows and calibration procedures.
Catalog No. 50-002-99321 Supplier Medchemexpress LLC Supplier No. HYW032349S50MG
1,2-Propylene-d6 carbonate is the deuterium-labeled analogue of 1,2-propylene carbonate used as an analytical tracer and internal standard for quantitative NMR, GC-MS, and LC-MS. It is supplied as a high-purity, isotopically labeled small-molecule reagent for method development, calibration, and quantitation workflows.
  • Deuterium-labeled (d6) isotopologue for mass and NMR differentiation.
  • High purity (98.65%) suitable for analytical applications.
  • Molecular weight 108.13 and chemical formula C4D6O3.
  • Intended use as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Available in small package sizes for analytical and method development.
  • Compatible with quantitative workflows and calibration procedures.