Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-Pyridyl)piperazine

Catalog No. 4081665ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-816-65ML 5 mL
40-816-625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-816-65ML Supplier Sigma Aldrich Supplier No. 4081665ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-(2-Pyridyl)piperazine is a piperazine derivative.1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC., It may be employed as reagent for the fluorometric determination of airborne diisocyantes.

Specifications

CAS 34803-66-2
Density 1.072 g/mL (at 25°C (literature))
Boiling Point 120°C to 122°C (2 mmHg)
Molecular Formula C9H13N3
Refractive Index n20/D 1.595 (literature)
Linear Formula C9H13N3
MDL Number MFCD00006216
Quantity 5 mL
Synonym 2-Piperazinopyridine
Molecular Weight (g/mol) 163.22
Percent Purity ≥99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.