Learn More
Medchemexpress LLC 1,2-pyrrolidinedicarboxylic acid, 4-[[[[2-[[(1,1-dimethylethoxy)carbonyl]amino]ethyl]amino]carb... | 187223-15-0 | MFCD07366898 | 99.7% | 539.58 | C28H33N3O8 | 1 G
SDP
Catalog No. 5000202060
Encompass_Preferred
Fmoc-Hyp(Bom)-OH is an Fmoc-protected hydroxyproline derivative with a Bom-protected hydroxy side chain, supplied as a high-purity solid for use in solid-phase peptide synthesis and linker construction for ADCs and PROTACs.
  • Fmoc-protected hydroxyproline with Bom side-chain protection.
  • High purity suitable for peptide synthesis and linker construction.
  • Molecular formula C28H33N3O8; molecular weight 539.58 g/mol.
  • Available in small lab-scale packages (1 G, 5 G, 10 G).
  • Characterized by HPLC purity 99.73%.
Catalog No. 50-002-02060 Supplier Medchemexpress LLC Supplier No. HY791251G
Fmoc-Hyp(Bom)-OH is an Fmoc-protected hydroxyproline derivative with a Bom-protected hydroxy side chain, supplied as a high-purity solid for use in solid-phase peptide synthesis and linker construction for ADCs and PROTACs.
  • Fmoc-protected hydroxyproline with Bom side-chain protection.
  • High purity suitable for peptide synthesis and linker construction.
  • Molecular formula C28H33N3O8; molecular weight 539.58 g/mol.
  • Available in small lab-scale packages (1 G, 5 G, 10 G).
  • Characterized by HPLC purity 99.73%.