Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-pyrrolidinedicarboxylic acid, 4-hydroxy-, 1-(1,1-dimethylethyl)-2-methyl ester (2S,4S) | 102195-79-9 | MFCD00237541 | >98.0% | 245.28 g/mol | C11H19NO5 | 100 G
SDP

Catalog No. 5000204023
Encompass_Preferred

N-Boc-cis-4-hydroxy-L-proline methyl ester is a Boc-protected hydroxyproline methyl ester used as a synthetic building block in organic synthesis and in linker development for conjugation chemistry. It is supplied as a white to off-white solid and is characterized by NMR and other analytical data for identity and purity.

  • Purity: ≥98.0% (NMR).
  • Molecular formula: C11H19NO5.
  • Molecular weight: 245.28 g/mol.
  • Appearance: white to off-white solid.
  • Water content (KF): 0.05%.
  • Optical rotation: [α]D (C = 1.00 g/100 mL EtOH) = -67.4°.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-04023 Supplier Medchemexpress LLC Supplier No. HYY0755100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Boc-cis-4-hydroxy-L-proline methyl ester is a Boc-protected hydroxyproline methyl ester used as a synthetic building block in organic synthesis and in linker development for conjugation chemistry. It is supplied as a white to off-white solid and is characterized by NMR and other analytical data for identity and purity.

  • Purity: ≥98.0% (NMR).
  • Molecular formula: C11H19NO5.
  • Molecular weight: 245.28 g/mol.
  • Appearance: white to off-white solid.
  • Water content (KF): 0.05%.
  • Optical rotation: [α]D (C = 1.00 g/100 mL EtOH) = -67.4°.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.