Learn More
Medchemexpress LLC 1,2-Pyrrolidinedicarboxylic acid, 5-oxo-, 1-(phenylmethyl) ester, (2S)- | 32159-21-0 | 99.9% | 263.25 | 25 G
SDP
Catalog No. 5000452733
Encompass_Preferred
Z-Pyr-OH (Cbz-L-pyroglutamic acid) can be used as a biochemical assay reagent. It appears as a white to off-white solid. The 1H NMR spectrum is consistent with its structure.
  • Can be stored for 3 years at room temperature
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
  • Tested and complies with specifications
  • For research use only
Catalog No. 50-004-52733 Supplier Medchemexpress LLC Supplier No. 25GHYW011750
Z-Pyr-OH (Cbz-L-pyroglutamic acid) can be used as a biochemical assay reagent. It appears as a white to off-white solid. The 1H NMR spectrum is consistent with its structure.
  • Can be stored for 3 years at room temperature
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
  • Tested and complies with specifications
  • For research use only