Learn More
Sonics & Materials Inc 1/2'' Replacement Tips
SDP
Catalog No. NC0311875
Encompass_Preferred
Standard ½”, ¾” and 1” probes are available with replaceable tips for use with water based samples. During use, tips erode and become less effective over time. A worn tip is easily removed and replaced.
Catalog No. NC0311875 Supplier Sonics & Materials Inc Supplier No. 6300406
Standard ½”, ¾” and 1” probes are available with replaceable tips for use with water based samples. During use, tips erode and become less effective over time. A worn tip is easily removed and replaced.