Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2'-Thienyl)ethylmercaptan 98%, Thermo Scientific™

Catalog No. AC265120025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC265120025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC265120025 Supplier Thermo Scientific Chemicals Supplier No. 265120025

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94089-02-8
Molecular Formula C6H8S2
Molecular Weight (g/mol) 144.25
InChI Key NCCCTQRHFVSOMP-UHFFFAOYSA-N
Synonym 1-2'-thienyl ethylmercaptan, 1-2-thienyl ethanethiol, 2-1-mercaptoethyl thiophene, 1-thiophen-2-yl ethane-1-thiol, 1-thiophen-2-yl ethanethiol, alpha-methylthiophene-2-methanethiol, 1-2-thienyl ethylmercaptan, 1-2'-thienyl ethyl mercaptan, alpha-methyl-2-thiophenemethanethiol, 2-thiophenemethanethiol, alpha-methyl
PubChem CID 2724626
IUPAC Name 1-thiophen-2-ylethanethiol
SMILES CC(C1=CC=CS1)S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.14
Infrared Spectrum Authentic
Refractive Index 1.5768 to 1.5786 (20°C, 589nm)
Quantity 2.5 g
Formula Weight 144.25
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1-(2'-Thienyl)ethylmercaptan, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement AIR SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.