Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[(2-Thienyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%

Catalog No. AC445580010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445580010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445580010 Supplier Thermo Scientific Chemicals Supplier No. 445580010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 864754-19-8 | C14H19BN2O2S | 290.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864754-19-8
Molecular Formula C14H19BN2O2S
Molecular Weight (g/mol) 290.19
MDL Number MFCD08706027
InChI Key ANSBFDYOIUZSNT-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-thienylmethyl-1h-pyrazole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-thiophen-2-ylmethyl-1h-pyrazole, 1-thiophen-2-ylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-thiophen-2-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-thiophen-2-ylmethyl-1h-pyrazole, 1-thiophen-2-ylmethyl pyrazole-4-boronic acid pinacol ester, 1-thiophen-2-ylmethyl-1h-pyrazol-4-boronic acid pinacol ester, 1-thiophen-2-ylmethyl pyrazole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-thiophen-2-ylmethyl pyrazole, pubchem18444
PubChem CID 17749925
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(thiophen-2-ylmethyl)pyrazole
SMILES CC1(C)OB(OC1(C)C)C1=CN(CC2=CC=CS2)N=C1

Specifications

Melting Point 57°C to 59°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 290.19
Percent Purity 97%
Chemical Name or Material 1-[(2-Thienyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.