Learn More
Chemglass Life Sciences 1/2" to 1/2" PFA Compression Fitting
SDP
Supplier:  Chemglass Life Sciences CG197521
Encompass_Preferred
1/2" to 1/2" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.
Catalog No. CG197521