Learn More
Chemglass Life Sciences PFA Compression Fitting, 1/2" to 3/4"
SDP
Catalog No. CG197522
Encompass_Preferred
1/2" to 3/4" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.
Catalog No. CG197522 Supplier Chemglass Life Sciences Supplier No. CG197522
1/2" to 3/4" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.