Learn More
Accuflow Systems Saint-Gobain Tri-Clamp Hose Barb Adapter, Polypropylene, 1/2 In, 25/PK
Catalog No. NC2323813
Encompass_Preferred
Tri-clamp to hose barb adapter with a smooth inner bore designed for sanitary fluid transfer and minimal dead volume. Polypropylene construction offers broad chemical compatibility and low extractables. Intended for 1/2 in (12.7 mm) tubing; packaged for bulk use in process and lab applications.
  • Polypropylene construction for chemical compatibility
  • Smooth inner bore (SIB) to minimize dead volume and entrapment
  • Tri-Clamp to hose barb configuration (barb x Tri-Clamp)
  • Tubing inner diameter: 1/2 in (12.7 mm)
  • Maximum operating pressure: 250 psi (typical, confirm for system conditions)
  • Pack quantity: 25 per pack for bulk assembly use
Catalog No. NC2323813 Supplier Accuflow Systems Supplier No. PFLS500PPAF
Tri-clamp to hose barb adapter with a smooth inner bore designed for sanitary fluid transfer and minimal dead volume. Polypropylene construction offers broad chemical compatibility and low extractables. Intended for 1/2 in (12.7 mm) tubing; packaged for bulk use in process and lab applications.
  • Polypropylene construction for chemical compatibility
  • Smooth inner bore (SIB) to minimize dead volume and entrapment
  • Tri-Clamp to hose barb configuration (barb x Tri-Clamp)
  • Tubing inner diameter: 1/2 in (12.7 mm)
  • Maximum operating pressure: 250 psi (typical, confirm for system conditions)
  • Pack quantity: 25 per pack for bulk assembly use