Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[2-(Trifluoromethyl)phenyl]imidazole 98.0+%, TCI America™
SDP

Catalog No. T29821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T29821G Supplier TCI America Supplier No. T29821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25371-96-4
Molecular Formula C10H7F3N2
Molecular Weight (g/mol) 212.175
MDL Number MFCD00041206
InChI Key WZBWBNCQUTXYEL-UHFFFAOYSA-N
Synonym trim, 1-2-trifluoromethyl phenyl imidazole, 1-2-trifluoromethylphenyl imidazole, 1-2-trifluoromethyl phenyl-1h-imidazole, 1h-imidazole,1-2-trifluoromethyl phenyl, dsstox_cid_25217, dsstox_rid_80756, dsstox_gsid_45217, 2-trifluoromethyl phenyl imidazole
PubChem CID 1359
IUPAC Name 1-[2-(trifluoromethyl)phenyl]imidazole
SMILES C1=CC=C(C(=C1)C(F)(F)F)N2C=CN=C2

Specifications

Melting Point 51°C
Color Yellow
Boiling Point 75°C
Quantity 1 g
Formula Weight 212.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[2-(Trifluoromethyl)phenyl]imidazole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.