Learn More
Holland Applied Technologies 1/2 USP CL VI EPDM SANITARY GS
Catalog No. NC9435567
Encompass_Preferred
0.5 in. USP CLASS VI EPDM GASKET, LOT NUMBER LASER ETCHED ON SIDE
This item is not returnable. View return policy
Catalog No. NC9435567 Supplier Holland Applied Technologies Supplier No. 42MPE0.5LE
0.5 in. USP CLASS VI EPDM GASKET, LOT NUMBER LASER ETCHED ON SIDE