Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Glass, Length x Dia. in mm: 12.7 x 9.52, Length x Dia. in inches: 1/2 x 3/8
SDP
Catalog No. CG200410
Encompass_Preferred
Stir Bar, Magnetic, Glass, Length x Dia. in mm: 12.7 x 9.52, Length x Dia. in inches: 1/2 x 3/8. Borosilicate covered Alnico magnet for use in high temperature reactions.
Catalog No. CG200410 Supplier Chemglass Life Sciences Supplier No. CG200410
Stir Bar, Magnetic, Glass, Length x Dia. in mm: 12.7 x 9.52, Length x Dia. in inches: 1/2 x 3/8. Borosilicate covered Alnico magnet for use in high temperature reactions.