Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc 1/2 YPD Plates, Case 100 x 25 mL
SDP

Catalog No. 507517594
Encompass_Preferred

YPD is a media variant derived from YPAD. YPAD consists of nutrient base (yeast extract and peptone) adenine supplement and a carbon source. YPDs formulation is the same but without the additional adenine.

YPD 1/2 Plates are useful for subculturing of untransformed yeast, yeast two-hybrid screening, yeast one-hybrid screening enrichment, and isolation of yeast; ade2 yeast will turn red on YPD.

Ready to use in petri dishes
Packed in 10 plates per sleeve
Re-closable zipper pouch for convenient storage

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Autoclave Sterilized

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7594 Supplier Quality Biological Inc Supplier No. 340184231
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YPD is a media variant derived from YPAD. YPAD consists of nutrient base (yeast extract and peptone) adenine supplement and a carbon source. YPDs formulation is the same but without the additional adenine.

YPD 1/2 Plates are useful for subculturing of untransformed yeast, yeast two-hybrid screening, yeast one-hybrid screening enrichment, and isolation of yeast; ade2 yeast will turn red on YPD.

Ready to use in petri dishes
Packed in 10 plates per sleeve
Re-closable zipper pouch for convenient storage

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Autoclave Sterilized

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.