Learn More
Andwin Scientific 1-200UL NS HR PIPET TIP 960/PK
SDP
Catalog No. NC0348190
Encompass_Preferred
The 1-200ul nonsterile polypropylene Pipet Tips feature flexible top that forms a tight seal with minimal ejection force, unbeveled point for easy sample dispensing and beveled mounting port for easy mounting for multichannel pipettors.
Catalog No. NC0348190 Supplier Andwin Scientific Supplier No. 37001528
The 1-200ul nonsterile polypropylene Pipet Tips feature flexible top that forms a tight seal with minimal ejection force, unbeveled point for easy sample dispensing and beveled mounting port for easy mounting for multichannel pipettors.