Learn More
Thermco Products Inc NIST Traceable Thermometer, -1/201C x 0.5C, Blue Spirit, 76mm Imm, 430mm long, Factory Certified @ 0, 25, 37, 44.5, 56, 105, 121, 180, 200C
Catalog No. NC0712314
Encompass_Preferred
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.
Catalog No. NC0712314 Supplier Thermco Products Inc Supplier No. ACC10073BLSFC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.