Learn More
MSC Ronstan Wire Rope Grip, 1/2 in Rope Dia, 316 Stainless Steel, Thimble
Catalog No. 5000534635
Encompass
This product is a Wire Rope Grip designed for a 1/2 in rope diameter. It is made from 316-grade stainless steel with an electropolished finish and features a thimble fitting type. A minimum of 3 clips are required for use. It is typically sold as a 3-piece kit.
  • Grip accessory type
  • For 1/2 in rope diameter
  • Stainless steel material
  • 316 material grade
  • Electropolished finish
  • Minimum of 3 clips required
  • Thimble fitting type
  • Sold as a 3-piece kit
Catalog No. 50-005-34635 Supplier MSC Supplier No. 73525511
This product is a Wire Rope Grip designed for a 1/2 in rope diameter. It is made from 316-grade stainless steel with an electropolished finish and features a thimble fitting type. A minimum of 3 clips are required for use. It is typically sold as a 3-piece kit.
  • Grip accessory type
  • For 1/2 in rope diameter
  • Stainless steel material
  • 316 material grade
  • Electropolished finish
  • Minimum of 3 clips required
  • Thimble fitting type
  • Sold as a 3-piece kit