Learn More
Medchemexpress LLC 1α,25-dihydroxy-3-epi-vitamin D3 | 61476-45-7 | 98.5% | 416.64 | C27H44O3 | 100 MG
SDP
Catalog No. 5000215019
Encompass_Preferred
(1S)-Calcitriol is the stereoisomeric form of calcitriol (1α,25-dihydroxyvitamin D3) used in biochemical and cellular research as a vitamin D receptor (VDR) ligand. It is intended for studies of VDR-mediated signaling pathways, regulation of keratinocyte proliferation, and parathyroid hormone suppression; supplied as a characterized research reagent in solid and solution formats.
  • Acts as a vitamin D receptor ligand for in vitro and biochemical assays.
  • High purity for reproducible experimental results.
  • Available in multiple solid mass sizes and DMSO solution formats.
  • Characterized by CAS number and molecular weight for traceability.
Catalog No. 50-002-15019 Supplier Medchemexpress LLC Supplier No. HY10002A100MG
Add to Cart
Add to Cart
Request a Quote
(1S)-Calcitriol is the stereoisomeric form of calcitriol (1α,25-dihydroxyvitamin D3) used in biochemical and cellular research as a vitamin D receptor (VDR) ligand. It is intended for studies of VDR-mediated signaling pathways, regulation of keratinocyte proliferation, and parathyroid hormone suppression; supplied as a characterized research reagent in solid and solution formats.
  • Acts as a vitamin D receptor ligand for in vitro and biochemical assays.
  • High purity for reproducible experimental results.
  • Available in multiple solid mass sizes and DMSO solution formats.
  • Characterized by CAS number and molecular weight for traceability.