Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1α,25-dihydroxy-3-epi-vitamin D3 | 61476-45-7 | MFCD28963706 | 98.5% | 416.64 | C27H44O3 | 25 MG
SDP

Catalog No. 5000215105
Encompass_Preferred

1α,25-dihydroxy-3-epi-vitamin D3 is a vitamin D3 metabolite and vitamin D receptor (VDR) agonist supplied as a solid research reagent for biochemical and cell-based studies. It is intended for laboratory use and should be protected from light.

  • CAS number 61476-45-7.
  • Molecular formula C27H44O3.
  • Molecular weight 416.64.
  • Purity 98.5%.
  • Appearance white to off-white solid.
  • Storage protect from light; solid: 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Solubility 100 mg/mL in DMSO (use freshly opened DMSO; ultrasonic assistance may be needed).
  • Typical pack sizes include 25 MG.

Catalog No. 50-002-15105 Supplier Medchemexpress LLC Supplier No. HY10002A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1α,25-dihydroxy-3-epi-vitamin D3 is a vitamin D3 metabolite and vitamin D receptor (VDR) agonist supplied as a solid research reagent for biochemical and cell-based studies. It is intended for laboratory use and should be protected from light.

  • CAS number 61476-45-7.
  • Molecular formula C27H44O3.
  • Molecular weight 416.64.
  • Purity 98.5%.
  • Appearance white to off-white solid.
  • Storage protect from light; solid: 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Solubility 100 mg/mL in DMSO (use freshly opened DMSO; ultrasonic assistance may be needed).
  • Typical pack sizes include 25 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.