Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1α,25-dihydroxy-3-epi-vitamin D3 | 61476-45-7 | MFCD28963706 | 98.5% | 416.64 | C27H44O3 | 50 MG
SDP

Catalog No. 5000215446
Encompass_Preferred

(1S)-Calcitriol (1α,25-dihydroxy-3-epi-vitamin D3) is a research-grade metabolite of vitamin D3 and a vitamin D receptor (VDR) agonist. Supplied as a white to off-white solid, it is used as an analytical standard and research reagent for studies of VDR-mediated biology, keratinocyte growth modulation, and calcium homeostasis.

  • Research-grade VDR agonist
  • High purity 98.5% (typical)
  • Molecular weight 416.64
  • Chemical formula C27H44O3
  • Supplied as 50 MG solid
  • Suitable for analytical and biological research

Catalog No. 50-002-15446 Supplier Medchemexpress LLC Supplier No. HY10002A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1S)-Calcitriol (1α,25-dihydroxy-3-epi-vitamin D3) is a research-grade metabolite of vitamin D3 and a vitamin D receptor (VDR) agonist. Supplied as a white to off-white solid, it is used as an analytical standard and research reagent for studies of VDR-mediated biology, keratinocyte growth modulation, and calcium homeostasis.

  • Research-grade VDR agonist
  • High purity 98.5% (typical)
  • Molecular weight 416.64
  • Chemical formula C27H44O3
  • Supplied as 50 MG solid
  • Suitable for analytical and biological research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.