Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1α,25-dihydroxy-3-epi-vitamin D3 | 61476-45-7 | MFCD28963706 | 98.5% | 416.64 g·mol⁻1 | C27H44O3 | 5 MG
SDP

Catalog No. 5000215020
Encompass_Preferred

1S-Calcitriol is a stereoisomer of calcitriol (1α,25-dihydroxyvitamin D3) that functions as a potent vitamin D receptor (VDR) agonist. It is provided as a high-purity solid research reagent for use in cellular, biochemical, and pharmacological studies of VDR signaling, calcium homeostasis, and related physiological processes.

  • Potent vitamin D receptor agonist.
  • Useful for studying keratinocyte proliferation and differentiation.
  • Supports investigation of parathyroid hormone regulation and calcium signaling.
  • Provided as a high-purity solid for analytical and biological assays.
  • Chemical formula C27H44O3; molecular weight 416.64 g·mol⁻1.

Catalog No. 50-002-15020 Supplier Medchemexpress LLC Supplier No. HY10002A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1S-Calcitriol is a stereoisomer of calcitriol (1α,25-dihydroxyvitamin D3) that functions as a potent vitamin D receptor (VDR) agonist. It is provided as a high-purity solid research reagent for use in cellular, biochemical, and pharmacological studies of VDR signaling, calcium homeostasis, and related physiological processes.

  • Potent vitamin D receptor agonist.
  • Useful for studying keratinocyte proliferation and differentiation.
  • Supports investigation of parathyroid hormone regulation and calcium signaling.
  • Provided as a high-purity solid for analytical and biological assays.
  • Chemical formula C27H44O3; molecular weight 416.64 g·mol⁻1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.