Learn More
Medchemexpress LLC 1α,25-dihydroxy-3-epi-vitamin D3 | 61476-45-7 | MFCD28963706 | 98.5% | 416.64 g/mol | C27H44O3 | 1 MG
SDP
Catalog No. 5000215482
Encompass_Preferred
(1S)-Calcitriol is the 1S stereoisomer of calcitriol, a biologically active vitamin D metabolite that functions as a vitamin D receptor (VDR) agonist. Supplied as a solid research reagent, it is intended for in vitro studies of vitamin D signaling and nuclear receptor biology. The product includes storage and solubility guidance to maintain compound integrity.
  • Acts as a vitamin D receptor (VDR) agonist for mechanistic studies.
  • High purity suitable for in vitro assays.
  • Recommended solubility in DMSO for stock preparations.
  • Stable as a solid when stored protected from light at 4°C.
  • Available in small solid pack sizes for research use.
Catalog No. 50-002-15482 Supplier Medchemexpress LLC Supplier No. HY10002A1MG
(1S)-Calcitriol is the 1S stereoisomer of calcitriol, a biologically active vitamin D metabolite that functions as a vitamin D receptor (VDR) agonist. Supplied as a solid research reagent, it is intended for in vitro studies of vitamin D signaling and nuclear receptor biology. The product includes storage and solubility guidance to maintain compound integrity.
  • Acts as a vitamin D receptor (VDR) agonist for mechanistic studies.
  • High purity suitable for in vitro assays.
  • Recommended solubility in DMSO for stock preparations.
  • Stable as a solid when stored protected from light at 4°C.
  • Available in small solid pack sizes for research use.