Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1α,25-dihydroxy-3-epi-vitamin D3 | 61476-45-7 | MFCD28963706 | 98.5% | 416.64 g·mol⁻¹ | C27H44O3 | 1 ML
SDP

Catalog No. 5000215669
Encompass_Preferred

(1S)-Calcitriol is the 3-epi stereoisomer of calcitriol (1α,25-dihydroxyvitamin D3), an active vitamin D metabolite used as a research reagent in VDR signaling, biochemical assays, and analytical standards. Supplied as a concentrated DMSO stock, it is intended for in vitro studies and should be handled protected from light and stored under recommended cold conditions.

  • Potent vitamin D receptor agonist for VDR-related assays and mechanistic studies.
  • Provided as a 10 mM solution in DMSO, 1 mL convenient stock format.
  • Chemical identity: 1α,25-dihydroxy-3-epi-vitamin D3 (CAS 61476-45-7).
  • Molecular weight 416.64 g·mol⁻¹.
  • Store protected from light; in solution keep at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-15669 Supplier Medchemexpress LLC Supplier No. HY10002A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1S)-Calcitriol is the 3-epi stereoisomer of calcitriol (1α,25-dihydroxyvitamin D3), an active vitamin D metabolite used as a research reagent in VDR signaling, biochemical assays, and analytical standards. Supplied as a concentrated DMSO stock, it is intended for in vitro studies and should be handled protected from light and stored under recommended cold conditions.

  • Potent vitamin D receptor agonist for VDR-related assays and mechanistic studies.
  • Provided as a 10 mM solution in DMSO, 1 mL convenient stock format.
  • Chemical identity: 1α,25-dihydroxy-3-epi-vitamin D3 (CAS 61476-45-7).
  • Molecular weight 416.64 g·mol⁻¹.
  • Store protected from light; in solution keep at -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.