Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1α,25-dihydroxy-3-epi-vitamin D3 | 61476-45-7 | MFCD28963706 | 98.5% | 416.64 g/mol | C27H44O3 | 10 MG
SDP

Catalog No. 5000215411
Encompass_Preferred

(1S)-Calcitriol is a research-grade vitamin D3 metabolite used to study vitamin D receptor signaling and related biological pathways. Supplied as a solid and in solution, it is intended for in vitro and in vivo research with storage and formulation guidance provided for experimental use.

  • High purity suitable for research applications (98.5%).
  • Molecular formula C27H44O3.
  • Molecular weight 416.64 g/mol.
  • CAS number 61476-45-7 for unambiguous identification.
  • Soluble in DMSO at 100 mg/mL; in vivo formulations documented.
  • Store protected from light; follow recommended cold storage for solutions and solids.

Catalog No. 50-002-15411 Supplier Medchemexpress LLC Supplier No. HY10002A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1S)-Calcitriol is a research-grade vitamin D3 metabolite used to study vitamin D receptor signaling and related biological pathways. Supplied as a solid and in solution, it is intended for in vitro and in vivo research with storage and formulation guidance provided for experimental use.

  • High purity suitable for research applications (98.5%).
  • Molecular formula C27H44O3.
  • Molecular weight 416.64 g/mol.
  • CAS number 61476-45-7 for unambiguous identification.
  • Soluble in DMSO at 100 mg/mL; in vivo formulations documented.
  • Store protected from light; follow recommended cold storage for solutions and solids.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.