Learn More
EUROIMMUN US INC 3P 1 25 DIHYDROXY VITAMIN D EIA
Catalog No. NC9874485
Encompass
1,25-Dihydroxy Vitamin D EIA, The 1,25-Dihydroxy Vitamin D EIA kit is a complete assay system intended for the purification of 1,25-dihydroxyvitamin D [1,25D] in human serum or plasma by immunopurification followed by quantification by enzymatic immunoassay to assess 1,25-dihydroxyvitamin D deficiency associated with renal disease. Certification: CE Marked; FDA Cleared Number of Tests: 96 (40 samples in duplicate) Clinical Area: Calcium Metabolism Sample Type: Serum. EDTA and Heparin Plasma Type: Manual Sample Volume: 0.5 mL with optional procedure for low sample volume Format: EIA Assay Range: 0 to 225 pg/mL (0 to 540.0 pmol/L)
This item is not returnable. View return policy
Catalog No. NC9874485 Supplier EUROIMMUN US INC 3P Supplier No. AC62F1
1,25-Dihydroxy Vitamin D EIA, The 1,25-Dihydroxy Vitamin D EIA kit is a complete assay system intended for the purification of 1,25-dihydroxyvitamin D [1,25D] in human serum or plasma by immunopurification followed by quantification by enzymatic immunoassay to assess 1,25-dihydroxyvitamin D deficiency associated with renal disease. Certification: CE Marked; FDA Cleared Number of Tests: 96 (40 samples in duplicate) Clinical Area: Calcium Metabolism Sample Type: Serum. EDTA and Heparin Plasma Type: Manual Sample Volume: 0.5 mL with optional procedure for low sample volume Format: EIA Assay Range: 0 to 225 pg/mL (0 to 540.0 pmol/L)