Learn More
Medchemexpress LLC 1,25-dihydroxyvitamin D3-13C3 (calcitriol-13C3) | 32222-06-3 | 97.0% | 413.57 | C24(13C3)H38O3 | 5 UG
SDP
Catalog No. 5000215447
Encompass_Preferred
Calcitriol-13C3 is a 13C-labeled form of calcitriol (1,25-dihydroxyvitamin D3) provided as a stable-isotope analytical standard for research use. It is intended as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS methods and is supplied as a small-volume ethanolic solution; protect from light and store under inert atmosphere for recommended shelf life.
Catalog No. 50-002-15447 Supplier Medchemexpress LLC Supplier No. HY10002S5UG
Calcitriol-13C3 is a 13C-labeled form of calcitriol (1,25-dihydroxyvitamin D3) provided as a stable-isotope analytical standard for research use. It is intended as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS methods and is supplied as a small-volume ethanolic solution; protect from light and store under inert atmosphere for recommended shelf life.