Learn More
Medchemexpress LLC 1β,25-dihydroxyvitamin D3 | 66791-71-7 | 98.0% | C27H44O3 | 10MG
SDP
Catalog No. 5000260720
Encompass_Preferred
Impurity B of Calcitriol (1β,25-dihydroxyvitamin D3) is an analytical reference standard supplied for laboratory research and analytical applications. It is an identified impurity of calcitriol, the hormonally active metabolite of vitamin D3 that modulates the vitamin D receptor. This material is for research use only and is not intended for human or clinical use.
  • High reported purity of 98.0% suitable for analytical assays.
  • Provides chemical identity C27H44O3 and molecular weight 416.64 g/mol.
  • Offered in multiple small package sizes for analytical workflows.
  • Intended specifically for research use only.
  • Supplied with a batch-specific certificate of analysis for traceability.
Catalog No. 50-002-60720 Supplier Medchemexpress LLC Supplier No. HY1329210MG
Impurity B of Calcitriol (1β,25-dihydroxyvitamin D3) is an analytical reference standard supplied for laboratory research and analytical applications. It is an identified impurity of calcitriol, the hormonally active metabolite of vitamin D3 that modulates the vitamin D receptor. This material is for research use only and is not intended for human or clinical use.
  • High reported purity of 98.0% suitable for analytical assays.
  • Provides chemical identity C27H44O3 and molecular weight 416.64 g/mol.
  • Offered in multiple small package sizes for analytical workflows.
  • Intended specifically for research use only.
  • Supplied with a batch-specific certificate of analysis for traceability.