Learn More
Uline Screw Top Pail - 1.25 Gallon, White Lid
Catalog No. NC1860533
Encompass_Preferred
Supplier has a minimum order quantity of 5. Quickly open and reseal for easy access. Vacuum tight ¿ Use with granulars, fine powders. Tamper evident with use of cable ties. Lid includes neoprene gasket for moisture-resistant storage. Molded, high-density polyethylene. FDA compliant.
Catalog No. NC1860533 Supplier Uline Supplier No. S18114W
Supplier has a minimum order quantity of 5. Quickly open and reseal for easy access. Vacuum tight ¿ Use with granulars, fine powders. Tamper evident with use of cable ties. Lid includes neoprene gasket for moisture-resistant storage. Molded, high-density polyethylene. FDA compliant.