Learn More
AirSea USA 1.25L Plain non-lacquer lined aluminium bottle with 45mm cap and tamper evident seal. UN Approved Single Packaging Aluminium Bottle – for the transportation of (Liquids) Dangerous Goods.
Catalog No. NC2101134
Encompass_Preferred
UN Mark: 1B1/X1.2/250, This aluminium bottle is UN approved as a single packaging for the transport of hazardous goods, but can also be used as an inner packaging within the approved 4G outer package.
This item is not returnable. View return policy
Catalog No. NC2101134 Supplier AirSea USA Supplier No. 182
UN Mark: 1B1/X1.2/250, This aluminium bottle is UN approved as a single packaging for the transport of hazardous goods, but can also be used as an inner packaging within the approved 4G outer package.