Learn More
BioVendor, LLC 1,25(OH)2 Vitamin D Extraction Cartridges (42 ct.)
SDP
Catalog No. 502069267
Encompass
Silica Extraction Cartridges for 1,25(OH)2 Vitamin D Total ELISA (42 ct.) For use with cat. no. RIS021R. Due to low levels and presence of similar metabolites, 1,25(OH)2 Vitamin D measurement requires extraction and separation by chromatography.
Catalog No. 50-206-9267 Supplier BioVendor, LLC Supplier No. 3019700
Silica Extraction Cartridges for 1,25(OH)2 Vitamin D Total ELISA (42 ct.) For use with cat. no. RIS021R. Due to low levels and presence of similar metabolites, 1,25(OH)2 Vitamin D measurement requires extraction and separation by chromatography.