Learn More
Cayman Chemical 1-(2-Amino-3-hydroxyphenyl)ethanone, Purity: ≥98%, CAS: 4502-10-7, 500 mg
SDP
Catalog No. 502913087
Encompass_Preferred
An active metabolite of 2-amino acetophenone; increases survival in a mouse model of P. aeruginosa-induced post-burn infection at 7.6 mg/kg
Catalog No. 50-291-3087 Supplier Cayman Chemical Supplier No. 36416500 MG
An active metabolite of 2-amino acetophenone; increases survival in a mouse model of P. aeruginosa-induced post-burn infection at 7.6 mg/kg