Learn More
Cayman Chemical 1,2-Dibutyryl-3-Palmitoyl-rac-glycerol, Purity: ≥98%, CAS: 74055-98-4, 50 mg
SDP
Catalog No. 502939123
Encompass_Preferred
A triacylglycerol; has been found in butterfat and maternal Dufour's glands of Anthophora bees
Catalog No. 50-293-9123 Supplier Cayman Chemical Supplier No. 2727950 MG
A triacylglycerol; has been found in butterfat and maternal Dufour's glands of Anthophora bees