Learn More
Cayman Chemical 1,2-Dierucoyl-sn-glycero-3-PC, Purity: ≥98%, CAS: 51779-95-4, 250 mg
SDP
Catalog No. 502936355
Encompass_Preferred
A phospholipid; has been used in the study of lipid membranes and to determine the effect of long-chain phospholipids on the secondary structure of hIAPP
Catalog No. 50-293-6355 Supplier Cayman Chemical Supplier No. 25728250 MG
A phospholipid; has been used in the study of lipid membranes and to determine the effect of long-chain phospholipids on the secondary structure of hIAPP