Learn More
Cayman Chemical 1,2-Dierucoyl-sn-glycero-3-PE, Purity: ≥98%, CAS: 904304-57-0, 10 mg
SDP
Catalog No. 502944758
Encompass_Preferred
A phospholipid; has been used in the generation of LNPs for the delivery of mRNA in vivo; has been used in the synthesis of various PEG-conjugates for incorporation into liposomes; LNPs containing 1,2-dierucoyl-sn-glycero-3-PE and encapsulating mRNA encoding hEPO increase serum hEPO levels in mice
Catalog No. 50-294-4758 Supplier Cayman Chemical Supplier No. 4109010 MG
A phospholipid; has been used in the generation of LNPs for the delivery of mRNA in vivo; has been used in the synthesis of various PEG-conjugates for incorporation into liposomes; LNPs containing 1,2-dierucoyl-sn-glycero-3-PE and encapsulating mRNA encoding hEPO increase serum hEPO levels in mice