Learn More
Cayman Chemical 1 2DIheptadecnoylsnglycer 10mg
SDP
Catalog No. 502918963
Encompass_Preferred
A glycerophospholipid; has been used as an internal standard for the detection of phosphatidylethanolamines in human adipose tissue
Catalog No. 50-291-8963 Supplier Cayman Chemical Supplier No. 3831910 MG
A glycerophospholipid; has been used as an internal standard for the detection of phosphatidylethanolamines in human adipose tissue