Learn More
Cayman Chemical 1,2-Dihexadecyl-sn-glycero-3-PC, Purity: ≥95%, CAS: 36314-47-3, 10 mg
SDP
Catalog No. 502936653
Encompass_Preferred
A phospholipid; commonly used in the generation of liposomes and artificial membranes to study membrane dynamics
Catalog No. 50-293-6653 Supplier Cayman Chemical Supplier No. 2558810 MG
A phospholipid; commonly used in the generation of liposomes and artificial membranes to study membrane dynamics