Learn More
Cayman Chemical 1 2DIlauroylracglycerol 10mg
SDP
Catalog No. 502932782
Encompass_Preferred
A diacylglycerol; has been used as an internal standard for the quantification of diacylglycerols in rat desheathed sciatic nerves; has been used in monomolecular films as substrates to measure the effects of surface pressure and pancreatic procolipase and colipase on the activity of porcine pancreatic lipase
Catalog No. 50-293-2782 Supplier Cayman Chemical Supplier No. 2684610 MG
A diacylglycerol; has been used as an internal standard for the quantification of diacylglycerols in rat desheathed sciatic nerves; has been used in monomolecular films as substrates to measure the effects of surface pressure and pancreatic procolipase and colipase on the activity of porcine pancreatic lipase